1-Bromo-3-butoxy-5-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

21960-500MG 21960-1G 21960-5G 21960-25G
77324-354EA 218.36 USD
77324-354 77348-248 77360-674 77384-088
1-Bromo-3-butoxy-5-(trifluoromethyl)benzene ≥97%
1-Bromo-3-butoxy-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrF₃O
MW: 297.112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28977075
CAS Number: 1866220-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR