5-Isopropoxy-2-methylphenol ≥97%

Supplier: AOBChem USA

Warning

21913-500MG 21913-1G 21913-5G 21913-25G
77324-350EA 212.18 USD
77324-350 77348-244 77360-670 77384-086
5-Isopropoxy-2-methylphenol ≥97%
5-Isopropoxy-2-methylphenol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD16997345
CAS Number: 918445-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR