(2,6-Difluoro-4-methylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

21823-500MG 21823-1G 21823-25G 21823-5G
77324-344EA 285.31 USD
77324-344 77348-242 77384-084 77372-526
(2,6-Difluoro-4-methylphenyl)methanol ≥97%
(2,6-Difluoro-4-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈F₂O
MW: 158.145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847294
CAS Number: 252004-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR