2-Bromo-5-(phenoxymethyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

21815-500MG 21815-1G 21815-5G 21815-25G
77324-342EA 210.12 USD
77324-342 77348-240 77360-664 77384-082
2-Bromo-5-(phenoxymethyl)pyridine ≥95%
2-Bromo-5-(phenoxymethyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrNO
MW: 264.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32706994
CAS Number: 845973-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR