Methyl 2,3-dimethoxy-6-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

21765-500MG 21765-1G 21765-5G 21765-25G
77324-336EA 196.73 USD
77324-336 77348-238 77360-658 77384-076
Methyl 2,3-dimethoxy-6-fluorobenzoate ≥97%
Methyl 2,3-dimethoxy-6-fluorobenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₄
Storage Temperature: Ambient
MDL Number: MFCD16038837
CAS Number: 1260793-27-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR