2,3-Dimethoxy-6-fluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

21763-500MG 21763-1G 21763-5G 21763-25G
77324-334EA 184.37 USD
77324-334 77347-148 77360-656 77384-074
2,3-Dimethoxy-6-fluorobenzyl alcohol ≥97%
2,3-Dimethoxy-6-fluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₁₁FO₃
Storage Temperature: Ambient
MDL Number: MFCD15528898
CAS Number: 1260814-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR