2,5-Difluoro-6-iodoanisole ≥97%

Supplier: AOBChem USA

Warning

21760-500MG 21760-1G 21760-5G 21760-25G
77324-332EA 161.71 USD
77324-332 77345-830 77360-654 77384-072
2,5-Difluoro-6-iodoanisole ≥97%
2,5-Difluoro-6-iodoanisole

Precaución: for Research Use Only

Formula: C₇H₅F₂IO
Storage Temperature: Ambient
MDL Number: MFCD12922646
CAS Number: 1208074-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR