6-Fluoro-2-methyl-3-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

21736-500MG 21736-1G 21736-5G 21736-25G
77324-324EA 145.23 USD
77324-324 77345-120 77360-648 77384-066
6-Fluoro-2-methyl-3-(methylthio)pyridine ≥97%
6-Fluoro-2-methyl-3-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₇H₈FNS
MW: 157.209 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20486345
CAS Number: 1204336-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR