4-Carboxy-2,6-difluorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

21735-500MG 21735-1G 21735-5G
77336-160EA 380.07 USD
77336-160 77348-234 77372-520
4-Carboxy-2,6-difluorophenylboronic acid pinacol ester ≥97%
4-Carboxy-2,6-difluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₅BF₂O₄
Storage Temperature: Ambient
MDL Number: MFCD22493663
CAS Number: 1031857-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR