3-Chloro-2-fluoro-4-(hydroxymethyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

21702-500MG 21702-1G 21702-5G 21702-25G
77324-322EA 251.32 USD
77324-322 77348-232 77360-646 77384-064
3-Chloro-2-fluoro-4-(hydroxymethyl)pyridine ≥97%
3-Chloro-2-fluoro-4-(hydroxymethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₅ClFNO
MW: 161.561 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12756633
CAS Number: 1149587-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR