5-Formyl-2-methylthiophen-3-yl-boronic acid ≥97%

Supplier: AOBChem USA

Warning

21614-500MG 21614-1G 21614-5G
77336-052EA 375.95 USD
77336-052 77348-226 77372-510
5-Formyl-2-methylthiophen-3-yl-boronic acid ≥97%
5-Formyl-2-methylthiophen-3-yl-boronic acid

Precaución: for Research Use Only

Formula: C₆H₇BO₃S
MW: 169.994 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12032184
CAS Number: 1310405-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR