6-Chloro-5-methylnicotinic acid isopropyl ester ≥97%

Supplier: AOBChem USA

Warning

21591-500MG 21591-1G 21591-5G 21591-25G
77324-320EA 141.11 USD
77324-320 77344-774 77360-644 77384-062
6-Chloro-5-methylnicotinic acid isopropyl ester ≥97%
6-Chloro-5-methylnicotinic acid isopropyl ester

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD26387896
CAS Number: 1122090-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR