3-Chloro-2,5-dimethoxypyridine ≥97%

Supplier: AOBChem USA

Warning

21573-500MG 21573-1G 21573-5G
77335-620EA 361.53 USD
77335-620 77348-222 77372-508
3-Chloro-2,5-dimethoxypyridine ≥97%
3-Chloro-2,5-dimethoxypyridine

Precaución: for Research Use Only

Formula: C₇H₈ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD16610312
CAS Number: 1227499-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR