5-Hydroxy-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

21531-1G 21531-5G 21531-25G
77341-590EA 94.76 USD
77341-590 77360-638 77372-504
5-Hydroxy-2-methylbenzaldehyde ≥97%
5-Hydroxy-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₈O₂
Storage Temperature: Refrigerator
MDL Number: MFCD16997527
CAS Number: 23942-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR