1-Bromo-3-[(3-methoxypheny)methoxy]benzene ≥97%

Supplier: AOBChem USA

Warning

21477-500MG 21477-1G 21477-5G 21477-25G
77324-314EA 161.71 USD
77324-314 77345-828 77360-636 77384-058
1-Bromo-3-[(3-methoxypheny)methoxy]benzene ≥97%
1-Bromo-3-[(3-methoxypheny)methoxy]benzene

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrO₂
Storage Temperature: Ambient
MDL Number: MFCD11127628
CAS Number: 1019446-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR