2,6-Dichloro-3-nitrophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

21456-500MG 21456-1G 21456-5G 21456-25G
77324-312EA 154.5 USD
77324-312 77345-534 77360-634 77384-056
2,6-Dichloro-3-nitrophenylboronic acid pinacol ester ≥97%
2,6-Dichloro-3-nitrophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₄BCl₂NO₄
Storage Temperature: Ambient
MDL Number: MFCD19105374
CAS Number: 1451391-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR