1-(Benzyloxy)-3-bromo-2-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

21403-1G 21403-5G 21403-25G
77341-184EA 74.16 USD
77341-184 77360-632 77372-502
1-(Benzyloxy)-3-bromo-2-fluorobenzene ≥97%
1-(Benzyloxy)-3-bromo-2-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrFO
Storage Temperature: Ambient
MDL Number: MFCD28977061
CAS Number: 295376-29-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR