(6-Chloro-2-fluoropyridin-3-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

21394-500MG 21394-1G 21394-5G 21394-25G
77324-310EA 147.29 USD
77324-310 77345-250 77360-630 77384-054
(6-Chloro-2-fluoropyridin-3-yl)methanol ≥97%
(6-Chloro-2-fluoropyridin-3-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₅ClFNO
Storage Temperature: Ambient
MDL Number: MFCD16610401
CAS Number: 1227509-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR