5-Bromo-2-fluoro-3-formylphenylboronic acid, pinacol ester ≥97%

Supplier: AOBChem USA

Warning

21326-500MG 21326-1G 21326-5G 21326-25G
77324-306EA 136.99 USD
77324-306 77344-578 77360-624 77384-050
5-Bromo-2-fluoro-3-formylphenylboronic acid, pinacol ester ≥97%
5-Bromo-2-fluoro-3-formylphenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₅BBrFO₃
MW: 328.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19105373
CAS Number: 1451391-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR