tert-Butyl 2-bromonicotinate ≥95%

Supplier: AOBChem USA

Warning

21295-500MG 21295-1G 21295-5G 21295-25G
77324-304EA 104.03 USD
77324-304 77343-068 77360-616 77372-492
tert-Butyl 2-bromonicotinate ≥95%
tert-Butyl 2-bromonicotinate

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrNO₂
MW: 258.112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07367920
CAS Number: 168629-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR