3-(3'-Methoxybenzyloxy)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 3-(3-Methoxybenzyloxy)phenylboronic acid

21235-500MG 21235-1G 21235-5G 21235-25G
77324-298EA 140.08 USD
77324-298 77344-688 77360-608 77384-044
3-(3'-Methoxybenzyloxy)phenylboronic acid ≥95%
3-(3'-Methoxybenzyloxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₅BO₄
MW: 258.077 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07783019
CAS Number: 871125-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR