2,6-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

21133-500MG 21133-1G 21133-5G 21133-25G
77324-292EA 222.48 USD
77324-292 77348-212 77360-602 77384-038
2,6-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-benzaldehyde ≥97%
2,6-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-benzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₅BCl₂O₃
MW: 300.973 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22494892
CAS Number: 1190989-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR