5-Bromo-2-methylsulfanylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

21121-500MG 21121-1G 21121-5G 21121-25G
77324-290EA 99.91 USD
77324-290 77342-752 77360-600 77372-488
5-Bromo-2-methylsulfanylbenzaldehyde ≥97%
5-Bromo-2-methylsulfanylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇BrOS
MW: 231.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20484745
CAS Number: 847978-59-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR