2-Bromo-4-chloro-5-(trifluoromethyl)benzenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-4-chloro-5-(trifluoromethyl)phenol

21101-1G 21101-5G 21101-25G
77341-218EA 77.25 USD
77341-218 77360-598 77372-486
2-Bromo-4-chloro-5-(trifluoromethyl)benzenol ≥97%
2-Bromo-4-chloro-5-(trifluoromethyl)benzenol

Precaución: for Research Use Only

Formula: C₇H₃BrClF₃O
MW: 275.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14560534
CAS Number: 1203898-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR