tert-Butyl(3,5-difluorophenoxy)dimethylsilane ≥97%

Supplier: AOBChem USA

Warning

21093-500MG 21093-1G 21093-5G 21093-25G
77324-288EA 120.51 USD
77324-288 77343-662 77360-596 77372-484
tert-Butyl(3,5-difluorophenoxy)dimethylsilane ≥97%
tert-Butyl(3,5-difluorophenoxy)dimethylsilane

Precaución: for Research Use Only

Formula: C₁₂H₁₈F₂OSi
MW: 244.353 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23164074
CAS Number: 917827-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR