1-Bromo-4-chloro-2-fluoro-3-propoxybenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
21004-500MG
21004-1G
21004-5G
21004-25G
77324-282EA
208.06
USD
77324-282
77348-206
77360-586
77384-032
1-Bromo-4-chloro-2-fluoro-3-propoxybenzene ≥97%
1-Bromo-4-chloro-2-fluoro-3-propoxybenzene
Precaución: for Research Use Only
Formula:
C₉H₉BrClFO MW: 267.523 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28977048 CAS Number: 1881295-73-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...