Methyl 2-chloro-6-fluoro-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

20990-500MG 20990-1G 20990-5G 20990-25G
77324-280EA 138.02 USD
77324-280 77344-576 77360-584 77384-030
Methyl 2-chloro-6-fluoro-3-methylbenzoate ≥97%
Methyl 2-chloro-6-fluoro-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₈ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD06203781
CAS Number: 773874-00-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR