3-Bromo-2,5,6-trifluorophenol ≥95%

Supplier: AOBChem USA

Warning

209842-500MG 209842-1G 209842-5G
77324-278EA 340.93 USD
77324-278 77348-204 77372-478
3-Bromo-2,5,6-trifluorophenol ≥95%
3-Bromo-2,5,6-trifluorophenol

Precaución: for Research Use Only

Formula: C₆H₂BrF₃O
MW: 226.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30529443
CAS Number: 1936039-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR