N-(3-Iodo-5-(trifluoromethyl)pyridin-2-yl)pivalamide ≥97%

Supplier: AOBChem USA

Warning

20947-500MG 20947-1G 20947-5G
77338-192EA 469.68 USD
77338-192 77348-202 77372-476
N-(3-Iodo-5-(trifluoromethyl)pyridin-2-yl)pivalamide ≥97%
N-(3-Iodo-5-(trifluoromethyl)pyridin-2-yl)pivalamide

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₃IN₂O
MW: 372.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17171313
CAS Number: 1002916-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR