2-Chloro-4-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

20897-500MG 20897-1G 20897-5G 20897-25G
77324-276EA 252.35 USD
77324-276 77348-200 77360-582 77384-028
2-Chloro-4-(methylthio)benzaldehyde ≥97%
2-Chloro-4-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇ClOS
Storage Temperature: Ambient
MDL Number: MFCD24676025
CAS Number: 146982-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR