1-Bromo-2-fluoro-3-(1-methylethoxy)-benzene ≥97%

Supplier: AOBChem USA

Warning

20836-500MG 20836-1G 20836-5G 20836-25G
77324-274EA 181.28 USD
77324-274 77346-774 77360-580 77384-026
1-Bromo-2-fluoro-3-(1-methylethoxy)-benzene ≥97%
1-Bromo-2-fluoro-3-(1-methylethoxy)-benzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO
MW: 233.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20526374
CAS Number: 1160293-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR