4-Methyl-3-benzyloxybenzoic acid methyl ester ≥97%

Supplier: AOBChem USA

Warning

20814-1G 20814-5G 20814-25G
77341-818EA 105.06 USD
77341-818 77360-578 77372-474
4-Methyl-3-benzyloxybenzoic acid methyl ester ≥97%
4-Methyl-3-benzyloxybenzoic acid methyl ester

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD06797834
CAS Number: 1017082-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR