4-Bromo-1-(cyclohexyloxy)-2-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

20800-500MG 20800-1G 20800-5G 20800-25G
77324-272EA 244.11 USD
77324-272 77348-196 77360-576 77384-024
4-Bromo-1-(cyclohexyloxy)-2-fluorobenzene ≥95%
4-Bromo-1-(cyclohexyloxy)-2-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₄BrFO
Storage Temperature: Ambient
MDL Number: MFCD28103576
CAS Number: 1369919-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR