3-Fluoro-4-propoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

20799-500MG 20799-1G 20799-5G 20799-25G
77324-270EA 166.86 USD
77324-270 77346-184 77360-574 77384-022
3-Fluoro-4-propoxybenzoic acid ≥97%
3-Fluoro-4-propoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
Storage Temperature: Ambient
MDL Number: MFCD01861238
CAS Number: 203115-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR