2-Bromo-6-(trifluoromethoxy)phenol ≥97%

Supplier: AOBChem USA

Warning

20753-500MG 20753-1G 20753-5G 20753-25G
77324-268EA 198.79 USD
77324-268 77348-194 77360-572 77384-020
2-Bromo-6-(trifluoromethoxy)phenol ≥97%
2-Bromo-6-(trifluoromethoxy)phenol

Precaución: for Research Use Only

Formula: C₇H₄BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD22414733
CAS Number: 1805029-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR