2-Bromo-1-chloro-4-methoxymethyl-benzene ≥97%

Supplier: AOBChem USA

Warning

20745-1G 20745-5G 20745-25G
77341-298EA 82.4 USD
77341-298 77360-570 77372-470
2-Bromo-1-chloro-4-methoxymethyl-benzene ≥97%
2-Bromo-1-chloro-4-methoxymethyl-benzene

Precaución: for Research Use Only

Formula: C₈H₈BrClO
MW: 235.506 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19105370
CAS Number: 1079402-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR