Methyl 6-fluoro-5-methoxynicotinate ≥97%

Supplier: AOBChem USA

Warning

20642-500MG 20642-1G
77340-538EA 1663.45 USD
77340-538 77348-188
Methyl 6-fluoro-5-methoxynicotinate ≥97%
Methyl 6-fluoro-5-methoxynicotinate

Precaución: for Research Use Only

Formula: C₈H₈FNO₃
Storage Temperature: Ambient
MDL Number: MFCD18258129
CAS Number: 1256791-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR