1,4-Dibromo-2-ethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1,4-Dibromo-2-ethoxy-benzene

20602-500MG 20602-1G 20602-5G 20602-25G
77324-264EA 199.82 USD
77324-264 77348-186 77360-566 77384-018
1,4-Dibromo-2-ethoxybenzene ≥97%
1,4-Dibromo-2-ethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₈Br₂O
MW: 279.957 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12547813
CAS Number: 1208077-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR