3,5-Dichloro(trifluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1,3-Dichloro-5-(trifluoromethoxy)benzene

205216-500MG 205216-1G 205216-5G 205216-25G
77324-260EA 235.87 USD
77324-260 77348-184 77360-562 77384-014
3,5-Dichloro(trifluoromethoxy)benzene ≥97%
3,5-Dichloro(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₃Cl₂F₃O
MW: 230.999 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28787555
CAS Number: 151276-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR