2-Fluoro-6-(propan-2-yloxy)phenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluro-6-isopropoxyphenol

20446-500MG 20446-1G 20446-5G 20446-25G
77324-256EA 234.84 USD
77324-256 77348-180 77360-558 77384-010
2-Fluoro-6-(propan-2-yloxy)phenol ≥97%
2-Fluoro-6-(propan-2-yloxy)phenol

Precaución: for Research Use Only

Formula: C₉H₁₁FO₂
MW: 170.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16997415
CAS Number: 1055315-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR