2,4-Dichloro-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

20441-500MG 20441-1G 20441-5G
77324-254EA 342.99 USD
77324-254 77348-178 77372-462
2,4-Dichloro-3-methoxybenzaldehyde ≥97%
2,4-Dichloro-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O₂
MW: 205.038 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28977031
CAS Number: 861787-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR