(4-Fluoro-3-isobutoxyphenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

20326-500MG 20326-1G 20326-5G
77335-522EA 357.41 USD
77335-522 77348-174 77372-454
(4-Fluoro-3-isobutoxyphenyl)boronic acid ≥95%
(4-Fluoro-3-isobutoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄BFO₃
MW: 212.026 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34755762
CAS Number: 2246729-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR