4-Bromo-1-fluoro-2-isobutoxybenzene ≥97%

Supplier: AOBChem USA

Warning

20325-500MG 20325-1G 20325-5G 20325-25G
77324-248EA 160.68 USD
77324-248 77345-806 77360-550 77384-006
4-Bromo-1-fluoro-2-isobutoxybenzene ≥97%
4-Bromo-1-fluoro-2-isobutoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrFO
Storage Temperature: Ambient
MDL Number: MFCD18917778
CAS Number: 1309933-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR