2,6-Difluoro-4-morpholin-4-yl-phenyl boronic acid ≥97%

Supplier: AOBChem USA

Warning

20302-500MG 20302-1G 20302-5G
77324-246EA 315.18 USD
77324-246 77348-172 77372-452
2,6-Difluoro-4-morpholin-4-yl-phenyl boronic acid ≥97%
2,6-Difluoro-4-morpholin-4-yl-phenyl boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂BF₂NO₃
MW: 243.015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16660036
CAS Number: 1093878-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR