Methyl 5-fluoro-2-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

20289-500MG 20289-1G 20289-5G 20289-25G
77324-244EA 154.5 USD
77324-244 77345-532 77360-548 77384-004
Methyl 5-fluoro-2-(methylthio)benzoate ≥97%
Methyl 5-fluoro-2-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₉H₉FO₂S
Storage Temperature: Ambient
MDL Number: MFCD29088119
CAS Number: 1879026-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR