2-Bromo-1,5-dichloro-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

20260-500MG 20260-1G 20260-5G 20260-25G
77324-240EA 252.35 USD
77324-240 77348-170 77360-544 77384-000
2-Bromo-1,5-dichloro-3-methylbenzene ≥97%
2-Bromo-1,5-dichloro-3-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂
MW: 239.925 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20233421
CAS Number: 876493-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR