3,5-Dichloro-4-propoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

20175-1G 20175-5G 20175-25G
77340-604EA 60.77 USD
77340-604 77360-540 77372-450
3,5-Dichloro-4-propoxybenzoic acid ≥97%
3,5-Dichloro-4-propoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀Cl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00662588
CAS Number: 41490-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR