(6-Isopropoxypyridin-3-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

20128-1G 20128-5G 20128-25G
77342-108EA 114.33 USD
77342-108 77360-538 77372-448
(6-Isopropoxypyridin-3-yl)methanol ≥95%
(6-Isopropoxypyridin-3-yl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD11110980
CAS Number: 1104461-69-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR