4-Bromo-3-methylphenylboronic acid N-butyldiethanolamine ester ≥97%

Supplier: AOBChem USA

Warning

19994-1G 19994-5G 19994-25G
77341-904EA 108.15 USD
77341-904 77360-534 77372-444
4-Bromo-3-methylphenylboronic acid N-butyldiethanolamine ester ≥97%
4-Bromo-3-methylphenylboronic acid N-butyldiethanolamine ester

Precaución: for Research Use Only

Formula: C₁₅H₂₃BBrNO₂
MW: 340.064 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19105365
CAS Number: 1451391-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR