2-Chloro-N-methoxy-N-methyl-6-(trifluoromethyl)benzamide ≥97%

Supplier: AOBChem USA

Warning

19992-1G 19992-5G 19992-25G
77340-602EA 60.77 USD
77340-602 77360-532 77372-442
2-Chloro-N-methoxy-N-methyl-6-(trifluoromethyl)benzamide ≥97%
2-Chloro-N-methoxy-N-methyl-6-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₁₀H₉ClF₃NO₂
MW: 267.632 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28977013
CAS Number: 2901066-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR